missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Trityl-L-cysteinamide, 98%

Manufacturer:  Alfa Aesar™ H6362806

 View more versions of this product

Catalog No. AAH6362806



Specifications

Specifications

S-Trityl-L-cysteinamide
5g
C22H22N2OS
h-cys trt-nh2, r-2-amino-3-tritylthio propanamide, s-trityl-l-cysteinamide, s-trityl-l-cysteine amide, s-trityl-l-cysteinamide hydrochloride, 2r-2-amino-3-triphenylmethyl sulfanyl propanamide, propanamide,2-amino-3-triphenylmethyl thio-, 2r, r-2-amino-3-tritylthio propanamide hydrochloride, ambotzhaa1560, h-cys trt-nh2.hcl
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)N)N
362.491
362.49
98%
White
166737-85-5
MFCD22126061
OHWBGKONMFYEKL-FQEVSTJZSA-N
(2R)-2-amino-3-tritylsulfanylpropanamide
44432703
Powder

Chemical Identifiers

166737-85-5
362.491
OHWBGKONMFYEKL-FQEVSTJZSA-N
44432703
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)N)N
C22H22N2OS
MFCD22126061
h-cys trt-nh2, r-2-amino-3-tritylthio propanamide, s-trityl-l-cysteinamide, s-trityl-l-cysteine amide, s-trityl-l-cysteinamide hydrochloride, 2r-2-amino-3-triphenylmethyl sulfanyl propanamide, propanamide,2-amino-3-triphenylmethyl thio-, 2r, r-2-amino-3-tritylthio propanamide hydrochloride, ambotzhaa1560, h-cys trt-nh2.hcl
(2R)-2-amino-3-tritylsulfanylpropanamide
SDS Documents