missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

s-Trioxane, 99+%, ACROS Organics™

Manufacturer:  Acros Organics 140290050

 View more versions of this product

Catalog No. AC140290050


 

Specifications

Specifications

s-Trioxane
45°C
0115.0°C
Colorless through White
5g
Authentic
≥99%
C3H6O3
1, 3, 5-Trioxacyclohexane, Trioxymethylene, 1, 3, 5-Trioxane
C1OCOCO1
90.078
CHEBI:38043
Crystals or Crystalline Flakes
1.1700g/mL
1.17
114.0°C to 116.0°C
61.0°C to 62.0°C
110-88-3
Glass bottle
99+%
MFCD00006563
BGJSXRVXTHVRSN-UHFFFAOYSA-N
1,3,5-trioxane
8081
90.08

Chemical Identifiers

110-88-3
90.078
BGJSXRVXTHVRSN-UHFFFAOYSA-N
8081
C3H6O3
MFCD00006563
1, 3, 5-Trioxacyclohexane, Trioxymethylene, 1, 3, 5-Trioxane
CHEBI:38043
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Suspected of damaging the unborn child.
Flammable solid.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents