Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Thiouronium chloride, polymer-supported, 0.8-1.0 mmol/g on Merrifield resin

Chemical Identifiers

Chemical Name or Material S-Thiouronium chloride
Name Note polymer-supported, 0.8 to 1.0mmol/g on Merrifield resin
MDL Number MFCD03458600
Synonym S-Methylisothiouronium choride on polystyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1937706

 
alfa aesar™
L1937706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note polymer-supported, 0.8 to 1.0mmol/g on Merrifield resin
Synonym S-Methylisothiouronium choride on polystyrene
Chemical Name or Material S-Thiouronium chloride
MDL Number MFCD03458600
Quantity 5g