missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-(Thiobenzoyl)thioglycolic Acid, TCI America™

Manufacturer:  TCI America T240125G

 View more versions of this product

Catalog No. T240125G



Specifications

Specifications

S-(Thiobenzoyl)thioglycolic Acid
128°C
942-91-6
MFCD00004927
XBEIANFIOZTEDE-UHFFFAOYSA-N
2-(benzenecarbonothioylsulfanyl)acetic acid
70336
Crystal-Powder at 20°C
Undesignated
25g
C9H8O2S2
(Thiobenzoylthio)acetic Acid
C1=CC=C(C=C1)C(=S)SCC(=O)O
212.281
212.28
≥97.0% (GC,T)

Chemical Identifiers

942-91-6
212.281
XBEIANFIOZTEDE-UHFFFAOYSA-N
70336
C1=CC=C(C=C1)C(=S)SCC(=O)O
C9H8O2S2
MFCD00004927
(Thiobenzoylthio)acetic Acid
2-(benzenecarbonothioylsulfanyl)acetic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents