Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-Tetrahydrofurfurylamine, 98+%

Chemical Identifiers

Chemical Name or Material (S)-(+)-Tetrahydrofurfurylamine
CAS 7175-81-7
Molecular Formula C5H11NO
Formula Weight 101.15g/mol
MDL Number MFCD00085303
UN Number UN2943
Beilstein 5725996
Synonym (S)-(+)-2-(Aminomethyl)tetrahydrofuran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1079003

 
alfa aesar™
L10790-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL10790MD

 
alfa aesar™
L10790-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 152° to 153°C
Flash Point 45°C (113°F)
Quantity 1g
Optical Rotation +12° (c=2 in Chloroform)
Chemical Name or Material (S)-(+)-Tetrahydrofurfurylamine
CAS 7175-81-7
Assay ≥98%
Molecular Formula C5H11NO
Formula Weight 101.15g/mol
MDL Number MFCD00085303
UN Number UN2943
Beilstein 5725996
Synonym (S)-(+)-2-(Aminomethyl)tetrahydrofuran
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAHYDROFURFURYLAMINE
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.