Please login to your online account to display your discounted pricing

(S)-(-)-Tetrahydro-2-furoic acid, 99% (98% e.e.), ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369510050

 View more versions of this product

Catalog No. AC369510050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 116.12
Quantity 5g
Color Clear colorless to pale yellow
Density 1.2100g/mL
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Packaging Glass bottle
Molecular Formula C5H8O3
CAS 87392-07-2
Name Note 98% e.e.
MDL Number MFCD00272639
Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
Specific Rotation -33° to -39° (20°C, 589nm) (c=1, CHCl3)
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
Name Note 98% e.e.
CAS 87392-07-2
Assay ≥98.5%
Molecular Formula C5H8O3
Formula Weight 116.12
MDL Number MFCD00272639
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents