Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-Tetrahydro-2-furoic acid, 98+%, ee 98%

Chemical Identifiers

Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
CAS 87392-07-2
Molecular Formula C5H8O3
Formula Weight 116.12
MDL Number MFCD00272639
UN Number UN3265
Beilstein 4658738
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1941103

 
alfa aesar™
L1941103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1941106

 
alfa aesar™
L1941106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1941114

 
alfa aesar™
L1941114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.12
Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
Assay ≥98%
MDL Number MFCD00272639
CAS 87392-07-2
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H8O3
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 244°C
Flash Point 139°C (282°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Assay Percent Notes ee 98%
Beilstein 4658738
Optical Rotation −4° (c=1 in Methanol)
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.