Please login to your online account to display your discounted pricing

(S)-tert-Leucinol, 95%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377532500

 View more versions of this product

Catalog No. AC377532500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Low Melting Mass or Liquid
Assay ≥94%
Formula Weight 117.19
Quantity 250mg
Color White or colorless
Chemical Name or Material (S)-tert-Leucinol
Assay Percent Range 95%
Molecular Formula C6H15NO
Linear Formula (CH3)3CCH(NH2)CH2OH
MDL Number MFCD00192250
CAS 112245-13-3
Packaging Glass bottle
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Specific Rotation +34° to +40° (25°C, 589nm) (c=1.5, EtOH)
CAS Min % 94.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-tert-Leucinol
CAS 112245-13-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H15NO
Linear Formula (CH3)3CCH(NH2)CH2OH
Formula Weight 117.19
MDL Number MFCD00192250
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents