Please login to your online account to display your discounted pricing

(S)-tert-Leucinol, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC377530010

 View more versions of this product

Catalog No. AC377530010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Low Melting Mass or Liquid
Assay ≥94%
Formula Weight 117.19
Quantity 1g
Color White or colorless
Density 90g/mL
Specific Rotation Condition + 37.50 (20.00°C c=2,EtOH)
Chemical Name or Material (S)-tert-Leucinol
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C6H15NO
Linear Formula (CH3)3CCH(NH2)CH2OH
MDL Number MFCD00192250
CAS 112245-13-3
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Specific Rotation +34° to +40° (25°C, 589nm) (c=1.5, EtOH)

Specifications

Chemical Name or Material (S)-tert-Leucinol
CAS 112245-13-3
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H15NO
Linear Formula (CH3)3CCH(NH2)CH2OH
Formula Weight 117.19
MDL Number MFCD00192250
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents