missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-tert-Butylsulfinamide, TCI America™

$67.22 - $245.98

Chemical Identifiers

CAS 343338-28-3
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.198
MDL Number MFCD05861480
InChI Key CESUXLKAADQNTB-ZETCQYMHSA-N
Synonym s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
PubChem CID 11355477
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29081G View Documents TCI America
B29081G
1g Each for $67.22
 
B29085G View Documents TCI America
B29085G
5g Each for $245.98
 
Specifications

Chemical Identifiers

343338-28-3
121.198
CESUXLKAADQNTB-ZETCQYMHSA-N
11355477
CC(C)(C)S(=O)N
C4H11NOS
MFCD05861480
s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
2-methylpropane-2-sulfinamide

Specifications

White-Yellow
1g
C4H11NOS
s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
CC(C)(C)S(=O)N
121.198
121.20
≥98.0% (GC)
106°C
343338-28-3
MFCD05861480
CESUXLKAADQNTB-ZETCQYMHSA-N
2-methylpropane-2-sulfinamide
11355477
Crystal-Powder at 20°C
(S)-(-)-tert-Butylsulfinamide
Safety and Handling

Safety and Handling

TSCA : No