missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-Styrene oxide, 94%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC449450050
Specifications
(S)-Styrene oxide | |
79°C | |
1.051 | |
-37.0°C | |
20780-54-5 | |
C8H8O | |
Solubility in water: practically insoluble | |
C1C(O1)C2=CC=CC=C2 | |
120.15 | |
CHEBI:51014 | |
94% |
1.0510g/mL | |
Glass bottle | |
192.0°C to 194.0°C | |
5g | |
92% min. (GC) | |
2s-2-phenyloxirane, ccris 4094, s-+-styrene oxide, s-2-phenyloxirane, s-epoxyethyl benzene, s-phenyloxirane, s-styrene oxide, styrene oxide, s, unii-av5p894c84 | |
AWMVMTVKBNGEAK-MRVPVSSYSA-N | |
(2S)-2-phenyloxirane | |
114946 | |
120.15 |
Chemical Identifiers
20780-54-5 | |
120.15 | |
2s-2-phenyloxirane, ccris 4094, s-+-styrene oxide, s-2-phenyloxirane, s-epoxyethyl benzene, s-phenyloxirane, s-styrene oxide, styrene oxide, s, unii-av5p894c84 | |
CHEBI:51014 | |
C1C(O1)C2=CC=CC=C2 |
C8H8O | |
AWMVMTVKBNGEAK-MRVPVSSYSA-N | |
114946 | |
(2S)-2-phenyloxirane |
Safety and Handling
GHS H Statement
May cause cancer.
Harmful in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger