missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-SNAP 5114, Tocris

GABA uptake inhibitor

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C30H35NO6
Formula Weight 505.61
Synonym 1-[2-[tris(4-methoxyphenyl)methoxy]ethyl]-(S)-3-piperidinecarboxylic acid
Chemical Name or Material (S)-SNAP 5114
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
156110 R & D Systems
RDS156110
10mg Each for $244.81
156150 R & D Systems
RDS156150
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>98%
505.61
(S)-SNAP 5114
C30H35NO6
1-[2-[tris(4-methoxyphenyl)methoxy]ethyl]-(S)-3-piperidinecarboxylic acid

Specifications

157604-55-2
C30H35NO6
1-[2-[tris(4-methoxyphenyl)methoxy]ethyl]-(S)-3-piperidinecarboxylic acid
(S)-SNAP 5114
>98%
505.61
10mg

Safety and Handling

Recommended Storage : Desiccate at Room Temperature