missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-SEGPHOS(regR), TCI America™

Manufacturer:  TCI America S09291G

 View more versions of this product

Catalog No. S09291G



Specifications

Specifications

(S)-(-)-SEGPHOS(regR)
217°C
210169-54-3
MFCD09753005
RZZDRSHFIVOQAF-UHFFFAOYSA-N
[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane
11017510
Crystal-Powder at 20°C
Undesignated
1g
C38H28O4P2
(S)-(-)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene)
C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8
610.586
610.59
≥99.0% (HPLC)

Chemical Identifiers

210169-54-3
610.586
RZZDRSHFIVOQAF-UHFFFAOYSA-N
11017510
C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8
C38H28O4P2
MFCD09753005
(S)-(-)-5,5′C-Bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis(diphenylphosphino)-3,3′C-bi(1,2-methylenedioxybenzene)
[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents