missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-sec-Phenethyl alcohol, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC217790050

 View more versions of this product

Catalog No. AC217790050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chiral derivatization reagent
Specifications

Specifications

(S)-(-)-sec-Phenethyl alcohol
98.5
1.0100g/mL
9°C to 11°C
85°C
Authentic
C6H5CH(CH3)OH
06, IV, 3029
1.5260 to 1.528
5g
WAPNOHKVXSQRPX-ZETCQYMHSA-N
C[C@H](O)C1=CC=CC=C1
(1S)-1-phenylethan-1-ol
122.17
CHEBI:16346
Liquid
1445-91-6
100.0
Colorless
98°C (20.0mmHg)
98.5% min. (GC)
C8H10O
MFCD00064264
Glass bottle
1-phenylethanol, s, 1s-1-phenylethan-1-ol, 1s-1-phenylethanol, 2mic4qly2x, s---1-phenylethanol, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol, s-1-phenylethanol, unii-2mic4qly2x
Solubility in water: 20g/L (20°C). Other solubilities: soluble in glycerol
1.01
−42.50 (20.00°C neat)
-42.5
443135
122.17
99%

Chemical Identifiers

1445-91-6
122.17
WAPNOHKVXSQRPX-ZETCQYMHSA-N
443135
(1S)-1-phenylethan-1-ol
C8H10O
MFCD00064264
1-phenylethanol, s, 1s-1-phenylethan-1-ol, 1s-1-phenylethanol, 2mic4qly2x, s---1-phenylethanol, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol, s-1-phenylethanol, unii-2mic4qly2x
CHEBI:16346
C[C@H](O)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only