Please login to your online account to display your discounted pricing

(S)-(+)-sec-Butanol, 99%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  218030050

 View more versions of this product

Catalog No. AC218030050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 74.12
Assay ≥98.5%
Quantity 5g
Color Clear colorless
Beilstein 01, 373
Merck Index 15, 1543
EINECS Number 224-168-1
TSCA TSCA
CAS Max % 100.0
Packaging Glass bottle
Synonym (S)-(+)-2-Butanol
Specific Rotation +12° to +14° (20°C, 589nm) (neat)
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Flammable liquid and vapor.
Name Note 99%
Molecular Formula C4H10O
Chemical Name or Material (S)-(+)-sec-Butanol
Linear Formula C2H5CH(OH)CH3
Assay Percent Range 99%
CAS Min % 98.5
CAS 4221-99-2
MDL Number MFCD00064281
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)

Specifications

Chemical Name or Material (S)-(+)-sec-Butanol
Name Note 99%
CAS 4221-99-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H10O
Linear Formula C2H5CH(OH)CH3
Formula Weight 74.12
MDL Number MFCD00064281
Synonym (S)-(+)-2-Butanol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents