Please login to your online account to display your discounted pricing

Alfa Aesar™ [(S,S)-TsDPEN-Ru(mesitylene)Cl]

Chemical Identifiers

Chemical Name or Material [(S,S)-TsDPEN-Ru(mesitylene)Cl]
CAS 174813-81-1
Formula Weight 622.18g/mol
MDL Number MFCD03791032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3799103

 
alfa aesar™
H37991-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Color Brown
Quantity 1g
Chemical Name or Material [(S,S)-TsDPEN-Ru(mesitylene)Cl]
CAS 174813-81-1
Physical Form Powder
Formula Weight 622.18g/mol
MDL Number MFCD03791032