Please login to your online account to display your discounted pricing

Alfa Aesar™ S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%


Quantity 25g

Manufacturer: alfa aesar™  L0057714

 View more versions of this product

Catalog No. AAL0057714

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 247.34
CAS 22236-45-9
Molecular Formula C9H13NO3S2
Chemical Name or Material S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
Assay 98%
MDL Number MFCD00013922
Melting Point 167° to 169°C
Beilstein 2119329
Quantity 25g
Synonym N-(p-Toluenesulfonyl)dimethylsulfoximine

Specifications

Chemical Name or Material S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
CAS 22236-45-9
Molecular Formula C9H13NO3S2
Formula Weight 247.34
MDL Number MFCD00013922
Beilstein 2119329
Synonym N-(p-Toluenesulfonyl)dimethylsulfoximine

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.