missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC293460050
Specifications
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine | |
Glass bottle | |
+8° (20°C c=3,EtOH) | |
Yellow | |
96% | |
95.0 | |
95% min. (GC) | |
MFCD00210016 | |
HDXYTIYVVNJFLU-YUMQZZPRSA-N | |
(2S,5S)-2,5-bis(methoxymethyl)pyrrolidine | |
7019262 | |
Liquid |
Authentic | |
1.4470 to 1.4490 | |
+ 8.00 | |
5g | |
93621-94-4 | |
100.0 | |
C8H17NO2 | |
2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine | |
COCC1CCC(N1)COC | |
159.23 | |
159.23 | |
96% |
Chemical Identifiers
93621-94-4 | |
159.23 | |
HDXYTIYVVNJFLU-YUMQZZPRSA-N | |
7019262 | |
COCC1CCC(N1)COC |
C8H17NO2 | |
MFCD00210016 | |
2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine | |
(2S,5S)-2,5-bis(methoxymethyl)pyrrolidine |
Safety and Handling
TSCA : TSCA