Please login to your online account to display your discounted pricing

Alfa Aesar™ (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine, 97%

Chemical Identifiers

Chemical Name or Material (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
CAS 163169-29-7
Formula Weight 481.35g/mol
MDL Number MFCD15144869
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63816MC

 
alfa aesar™
H63816-MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63816ME

 
alfa aesar™
H63816-ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Orange
Melting Point 158 to 162°C
Quantity 100mg
Sensitivity Air Sensitive
Chemical Name or Material (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
CAS 163169-29-7
Physical Form Solid
Assay 97%
Formula Weight 481.35g/mol
MDL Number MFCD15144869