missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S,S)-(-)-1,2-Diphenyl-1,2-ethanediol, +98%, ACROS Organics™

Chemical Identifiers

CAS 2325-10-2
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064255
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
Synonym 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethanediol, 1s,2s-1,2-diphenylethane-1,2-diol, hydrobenzoin,-, isohydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, s,s---hydrobenzoin, s,s-1,2-diphenylethylene glycol, s,s-hydrobenzoin, unii-wx45q7714b
PubChem CID 853020
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC279515000
SDS View Documents
Thermo Fisher Scientific
279515000
500mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2325-10-2
214.26
IHPDTPWNFBQHEB-KBPBESRZSA-N
853020
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
C14H14O2
MFCD00064255
1,2-ethanediol, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethanediol, 1s,2s-1,2-diphenylethane-1,2-diol, hydrobenzoin,-, isohydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, s,s---hydrobenzoin, s,s-1,2-diphenylethylene glycol, s,s-hydrobenzoin, unii-wx45q7714b
(1S,2S)-1,2-diphenylethane-1,2-diol

Specifications

Authentic
− 93.00 (20.00°C c=2.5,ethanol)
White
500mg
98.0
98+%
MFCD00064255
IHPDTPWNFBQHEB-KBPBESRZSA-N
(1S,2S)-1,2-diphenylethane-1,2-diol
853020
Crystalline Powder
(S,S)-(-)-1,2-Diphenyl-1,2-ethanediol, 98+%
Glass bottle
-92° to -94° (20°C, 589nm) (c=2.5, EtOH)
147°C to 150°C
2325-10-2
100.0
C14H14O2
1,2-ethanediol, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethanediol, 1s,2s-1,2-diphenylethane-1,2-diol, hydrobenzoin,-, isohydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, s,s---hydrobenzoin, s,s-1,2-diphenylethylene glycol, s,s-hydrobenzoin, unii-wx45q7714b
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
214.26
214.26
≥98.0%
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS