Please login to your online account to display your discounted pricing

Alfa Aesar™ (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$215.47 - $215.47

Chemical Identifiers

Chemical Name or Material (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C22H20N2·2HCl
Formula Weight 385.35
MDL Number MFCD09265324
Synonym (S,S)-1,2-Bis(1-naphthyl)-1,2-ethanediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2599303

  SDS
alfa aesar™
H2599303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH25993MD

  SDS
alfa aesar™
H25993MD
250mg Each for $215.47
Description & Specifications

Specifications

Formula Weight 385.35
Assay Percent Notes ee 99%
Health Hazard 1 Warning
Quantity 1g
MDL Number MFCD09265324
Chemical Name or Material (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C22H20N2·2HCl
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym (S,S)-1,2-Bis(1-naphthyl)-1,2-ethanediamine dihydrochloride
Assay 95%
Melting Point >200°C
Optical Rotation 235° (c=1 in Water)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.