Please login to your online account to display your discounted pricing

Alfa Aesar™ (S,S)-(-)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$200.41 - $566.54

Chemical Identifiers

Chemical Name or Material (S,S)-(-)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C14H6F10N2·2HCl
Formula Weight 465.13
MDL Number MFCD09265323
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2598103

  SDS
alfa aesar™
H2598103
1g Each for $566.54

AAH25981MD

  SDS
alfa aesar™
H25981MD
250mg Each for $200.41
Description & Specifications

Specifications

Formula Weight 465.13
Molecular Formula C14H6F10N2·2HCl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Notes ee 99%
MDL Number MFCD09265323
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (S,S)-(-)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride
Melting Point >200°C
Optical Rotation −28° (c=1 in Methanol)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.