Please login to your online account to display your discounted pricing

(S)-(+)-Rolipram, Tocris


PDE4 inhibitor. Less active enantiomer of rolipram

Quantity 50mg

Manufacturer: r & d systems  RDS135050

 View more versions of this product

Catalog No. 135050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 275.35
Quantity 50mg
Chemical Name or Material (S)-(+)-Rolipram
Assay Percent Range >99%
Molecular Formula C16H21NO3
CAS 85416-73-5
Synonym (4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (S)-(+)-Rolipram
Assay Percent Range >99%
Molecular Formula C16H21NO3
Formula Weight 275.35
Synonym (4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Accessories

Store at Room Temperature