Please login to your online account to display your discounted pricing

(S)-(+)-Rolipram, Tocris

PDE4 inhibitor. Less active enantiomer of rolipram

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material (S)-(+)-Rolipram
Assay Percent Range >99%
Molecular Formula C16H21NO3
Formula Weight 275.35
Synonym (4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

135010

 
r & d systems
RDS135010
10mg Each for $252.39

135050

 
r & d systems
RDS135050
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 275.35
Quantity 10mg
Chemical Name or Material (S)-(+)-Rolipram
Assay Percent Range >99%
Molecular Formula C16H21NO3
CAS 85416-73-5
Synonym (4S)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature