missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-Propylene Carbonate 98.0+%, TCI America™
Supplier: TCI America P148625G
Specifications
(S)-Propylene Carbonate | |
Yellow | |
C4H6O3 | |
25 g | |
RUOJZAUFBMNUDX-VKHMYHEASA-N | |
(4S)-4-methyl-1,3-dioxolan-2-one | |
2734444 | |
≥98.0% (GC) |
51260-39-0 | |
105°C | |
MFCD00798264 | |
(S)-4-Methyl-1,3-dioxolan-2-one | |
CC1COC(=O)O1 | |
102.089 | |
102.09 | |
Liquid |
Chemical Identifiers
51260-39-0 | |
102.089 | |
RUOJZAUFBMNUDX-VKHMYHEASA-N | |
2734444 | |
CC1COC(=O)O1 |
C4H6O3 | |
MFCD00798264 | |
(S)-4-Methyl-1,3-dioxolan-2-one | |
(4S)-4-methyl-1,3-dioxolan-2-one |
Safety and Handling
EINECSNumber : (5)-0524
RTECSNumber : FF9650000
TSCA : No