missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-α-Propargylglycine, 98%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441215000

 View more versions of this product

Catalog No. AC441215000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-α-Propargylglycine
Authentic
-32
>180°C
98% ee
97.5
97.5 to 102.5%
2s-2-amino-4-pentynoic acid, 2s-2-aminopent-4-ynoic acid, 4-pentynoic acid, 2-amino-, 2s, 8r9u8rvt6v, l-2-propynylglycine, l-propargylglycine, s-2-amino-4-pentynoic acid, s-2-aminopent-4-ynoic acid, s-propargylglycine, unii-8r9u8rvt6v
C#CCC(C(=O)O)N
113.12
113.12
98%,98% ee
98.0% min.
Glass Bottle
White to Yellow
500mg
23235-01-0
100.0
C5H7NO2
DGYHPLMPMRKMPD-BYPYZUCNSA-N
(2S)-2-aminopent-4-ynoic acid
168091
Powder

Chemical Identifiers

23235-01-0
113.12
2s-2-amino-4-pentynoic acid, 2s-2-aminopent-4-ynoic acid, 4-pentynoic acid, 2-amino-, 2s, 8r9u8rvt6v, l-2-propynylglycine, l-propargylglycine, s-2-amino-4-pentynoic acid, s-2-aminopent-4-ynoic acid, s-propargylglycine, unii-8r9u8rvt6v
(2S)-2-aminopent-4-ynoic acid
C5H7NO2
DGYHPLMPMRKMPD-BYPYZUCNSA-N
168091
C#CCC(C(=O)O)N
SDS Documents