Please login to your online account to display your discounted pricing

(S)-α-Propargylglycine, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-α-Propargylglycine
Name Note 98% ee
CAS 23235-01-0
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C5H7NO2
Formula Weight 113.12
Synonym (S)-2-Amino-4-pentynoic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC441211000

 
thermo fisher scientific
AC441211000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441215000

 
thermo fisher scientific
AC441215000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 113.12
Quantity 100mg
Color White to pale yellow
Assay 97.5 to 102.5%
CAS Max % 100.0
Chemical Name or Material (S)-α-Propargylglycine
Enantiomeric Excess 98.0% min.
Name Note 98% ee
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H7NO2
Packaging Glass bottle
CAS 23235-01-0
Synonym (S)-2-Amino-4-pentynoic acid
Specific Rotation -31° to -35° (20°C, 589nm) (c=1 in water)
CAS Min % 97.5