Learn More
(S)-(+)-Prolinol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0977914
Description
It is an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an active pharmaceutical intermediate.
Solubility
Fully miscible in water. Soluble in chloroform.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
(S)-(+)-Prolinol | |
1.4853 | |
74°C to 76°C (2mmHg) | |
25 g | |
+31° (c=1 in Toluene) | |
C5H11NO | |
79843 | |
Fully miscible in water. Soluble in chloroform. | |
C1CC(NC1)CO | |
101.149 | |
101.15 |
23356-96-9 | |
1.025 | |
42°C to 44°C | |
86°C (186°F) | |
Air sensitive; Hygroscopic | |
MFCD00005255 | |
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol | |
HVVNJUAVDAZWCB-YFKPBYRVSA-N | |
[(2S)-pyrrolidin-2-yl]methanol | |
640091 | |
98% |
Chemical Identifiers
23356-96-9 | |
101.149 | |
HVVNJUAVDAZWCB-YFKPBYRVSA-N | |
640091 | |
C1CC(NC1)CO |
C5H11NO | |
MFCD00005255 | |
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol | |
[(2S)-pyrrolidin-2-yl]methanol |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 245-605-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only