Please login to your online account to display your discounted pricing

(S)-(-)-Pindolol, Tocris


β3 partial agonist; More active enantiomer of pindolol

Quantity 10mg

Manufacturer: r & d systems  106010

 View more versions of this product

Catalog No. 106010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 248.32
Quantity 10mg
Chemical Name or Material (S)-(-)-Pindolol
Assay Percent Range >99%
Synonym (S)-1-(1H-indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol
Molecular Formula C14H20N2O2
CAS 26328-11-0
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material (S)-(-)-Pindolol
Assay Percent Range >99%
Molecular Formula C14H20N2O2
Formula Weight 248.32
Synonym (S)-1-(1H-indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol
Accessories

Desiccate at +4°C