Please login to your online account to display your discounted pricing

(S)-(-)-Pindolol, Tocris

β3 partial agonist; More active enantiomer of pindolol

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material (S)-(-)-Pindolol
Assay Percent Range >99%
Molecular Formula C14H20N2O2
Formula Weight 248.32
Synonym (S)-1-(1H-indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

106010

 
r & d systems
106010
10mg Each for $252.39

106050

 
r & d systems
106050
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 248.32
Quantity 10mg
Synonym (S)-1-(1H-indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol
Assay Percent Range >99%
Chemical Name or Material (S)-(-)-Pindolol
CAS 26328-11-0
Molecular Formula C14H20N2O2
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C