missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5517506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
S-Phenyl thioacetate | |
Colorless | |
99°C to 100°C (6 mmHg) | |
C8H8OS | |
CH3COSC6H5 | |
5 g | |
WBISVCLTLBMTDS-UHFFFAOYSA-N | |
S-phenyl ethanethioate | |
13630 | |
98% |
934-87-2 | |
1.124 g/mL | |
79°C (174°F) | |
1.57 | |
MFCD00008752 | |
s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl | |
CC(=O)SC1=CC=CC=C1 | |
152.211 | |
152.21 | |
Liquid |
Chemical Identifiers
934-87-2 | |
152.211 | |
WBISVCLTLBMTDS-UHFFFAOYSA-N | |
13630 | |
CC(=O)SC1=CC=CC=C1 |
C8H8OS | |
MFCD00008752 | |
s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl | |
S-phenyl ethanethioate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 213-294-2
RTECSNumber : AJ7559000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only