missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Phenyl thioacetate, 98%

Manufacturer:  Alfa Aesar™ H5517514

 View more versions of this product

Catalog No. AAH5517514



Specifications

Specifications

S-Phenyl thioacetate
79°C (174°F)
99°C to 100°C (6mmHg)
25g
C8H8OS
MFCD00008752
WBISVCLTLBMTDS-UHFFFAOYSA-N
S-phenyl ethanethioate
13630
Liquid
1.124 g/mL
1.57
Colorless
934-87-2
CH3COSC6H5
1-phenylsulfanylethanone
CC(=O)SC1=CC=CC=C1
152.211
152.21
98%

Chemical Identifiers

934-87-2
152.211
WBISVCLTLBMTDS-UHFFFAOYSA-N
13630
CC(=O)SC1=CC=CC=C1
C8H8OS
MFCD00008752
1-phenylsulfanylethanone
S-phenyl ethanethioate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 213-294-2

RTECSNumber : AJ7559000

TSCA : No

Recommended Storage : Ambient temperatures

Documents