Please login to your online account to display your discounted pricing

(S)-Phenyl superquat, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  337812500

 View more versions of this product

Catalog No. AC337812500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.23
Quantity 250mg
TSCA TSCA
CAS Max % 100.0
Packaging Glass bottle
Synonym (S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone
CAS Min % 97.5
Assay Percent Range 98%
Chemical Name or Material (S)-Phenyl superquat
CAS 168297-84-5
MDL Number MFCD00274262
Name Note 98%
Molecular Formula C11H13NO2

Specifications

Chemical Name or Material (S)-Phenyl superquat
Name Note 98%
CAS 168297-84-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H13NO2
Formula Weight 191.23
MDL Number MFCD00274262
Synonym (S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone