missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-Phenyl superquat, 98%, ACROS Organics™

Chemical Identifiers

CAS 168297-84-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C11H13NO2
Formula Weight 191.23
MDL Number MFCD00274262
Synonym (S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone
Chemical Name or Material (S)-Phenyl superquat, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC337812500 SDS Thermo Fisher Scientific
337812500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC337810010 SDS Thermo Fisher Scientific
337810010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

168297-84-5
100.0
C11H13NO2
MFCD00274262
(S)-Phenyl superquat, 98%
97.5
98%
191.23
(S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone

Specifications

191.23
100.0
(S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone
98%
MFCD00274262
(S)-Phenyl superquat, 98%
250mg
Glass bottle
97.5
168297-84-5
C11H13NO2

Safety and Handling

TSCA : TSCA