missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Phenyl-L-cysteine, 97%

Manufacturer:  Alfa Aesar™ H5503114

 View more versions of this product

Catalog No. AAH5503114



Specifications

Specifications

S-Phenyl-L-cysteine
C9H11NO2S
2268203
XYUBQWNJDIAEES-QMMMGPOBSA-N
(2R)-2-amino-3-phenylsulfanylpropanoic acid
119462
97%
25g
34317-61-8
MFCD01318758
2r-2-amino-3-phenylsulfanylpropanoic acid
C1=CC=C(C=C1)SCC(C(=O)O)N
197.252
197.25
200°C (decomposition)
+10° (c=1.5 in 1M NaOH)

Chemical Identifiers

34317-61-8
197.252
XYUBQWNJDIAEES-QMMMGPOBSA-N
119462
C1=CC=C(C=C1)SCC(C(=O)O)N
C9H11NO2S
MFCD01318758
2r-2-amino-3-phenylsulfanylpropanoic acid
(2R)-2-amino-3-phenylsulfanylpropanoic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents