Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-Phanephos, 98%, ee 98%

$211.22 - $211.22

Chemical Identifiers

Chemical Name or Material (S)-Phanephos
CAS 192463-40-4
Molecular Formula C40H34P2
Formula Weight 576.65g/mol
MDL Number MFCD03094575
Synonym (S)-(+)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25813MD

 
alfa aesar™
H25813-MD
250mg Each for $211.22
Description & Specifications

Specifications

Melting Point 226° to 230°C
Quantity 250mg
Chemical Name or Material (S)-Phanephos
CAS 192463-40-4
Assay 98%
Assay Percent Notes ee 98%
Molecular Formula C40H34P2
Formula Weight 576.65g/mol
MDL Number MFCD03094575
Synonym (S)-(+)-4,12-Bis(diphenylphosphino)[2.2]paracyclophane