missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-p-Tolyl p-Toluenesulfonothioate 98.0+%, TCI America™
Supplier: TCI America T31735G
Specifications
S-p-Tolyl p-Toluenesulfonothioate | |
74°C | |
C14H14O2S2 | |
5 g | |
HSAROCRPZOYKGS-UHFFFAOYSA-N | |
1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene | |
257647 | |
≥98.0% (HPLC) |
2943-42-2 | |
White | |
MFCD00092126 | |
p-Toluenesulfonothioic Acid S-p-Tolyl Ester | |
CC1=CC=C(SS(=O)(=O)C2=CC=C(C)C=C2)C=C1 | |
278.38 | |
278.38 | |
Crystalline Powder |
Chemical Identifiers
2943-42-2 | |
278.38 | |
HSAROCRPZOYKGS-UHFFFAOYSA-N | |
257647 | |
CC1=CC=C(SS(=O)(=O)C2=CC=C(C)C=C2)C=C1 |
C14H14O2S2 | |
MFCD00092126 | |
p-Toluenesulfonothioic Acid S-p-Tolyl Ester | |
1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene |
Safety and Handling
TSCA : No