missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-Nipecotic acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436540100

 View more versions of this product

Catalog No. AC436540100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(S)-(+)-Nipecotic acid
+ 4.6
247°C to 249°C
59045-82-8
C6H11NO2
XJLSEXAGTJCILF-YFKPBYRVSA-N
(3S)-piperidine-3-carboxylic acid
6575389
129.16
96%
Authentic
White
10g
96%
+-hexahydronicotinic acid
C1CC(CNC1)C(=O)O
129.16
CHEBI:222169
Powder

Chemical Identifiers

59045-82-8
129.16
+-hexahydronicotinic acid
CHEBI:222169
C1CC(CNC1)C(=O)O
C6H11NO2
XJLSEXAGTJCILF-YFKPBYRVSA-N
6575389
(3S)-piperidine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS Documents