Please login to your online account to display your discounted pricing

(S)-(+)-Niguldipine hydrochloride, Tocris

α1 antagonist, L-type Ca2+ channel blocker

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material (S)-(+)-Niguldipine hydrochloride
Assay Percent Range >98%
Molecular Formula C36H39N3O6·HCl
Formula Weight 646.18
Synonym (S)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

112350

 
r & d systems
RDS112350
50mg Each for $891.94

112310

 
r & d systems
RDS112310
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 646.18
Quantity 50mg
Chemical Name or Material (S)-(+)-Niguldipine hydrochloride
CAS 113145-69-0
Assay Percent Range >98%
Synonym (S)-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid, 3-(4,4-diphenyl-1-piperidinyl)propyl methyl ester hydrochloride
Molecular Formula C36H39N3O6·HCl
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C