missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-Nicotine, 99%

Manufacturer:  Alfa Aesar A1239814

 View more versions of this product

Catalog No. AAA1239814


 

Description

Description

Solubility
Miscible with water, alcohol, ether and light petroleum.

Notes
Hygroscopic. Air and light sensitive.
Specifications

Specifications

(S)-(-)-Nicotine
25g
99%
C10H14N2
UN1654
14,6524
CN1CCCC1C2=CN=CC=C2
162.236
CHEBI:17688
Liquid
1.017
54-11-5
99%
MFCD00006369
82109
SNICXCGAKADSCV-JTQLQIEISA-N
3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
89594
162.24

Chemical Identifiers

54-11-5
162.236
SNICXCGAKADSCV-JTQLQIEISA-N
CHEBI:17688
C10H14N2
MFCD00006369
89594
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310

GHS P Statement
P262-P280-P301+P310-P361-P302+P350-P310-P363-P321-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NICOTINE

RTECSNumber : QS5250000

TSCA : TSCA

storageNote1 : Air Sensitive and Hygroscopic

SDS Documents