Please login to your online account to display your discounted pricing

Alfa Aesar™ S-n-Propyl thioacetate, 98+%

Chemical Identifiers

Chemical Name or Material S-n-Propyl thioacetate
CAS 2307-10-0
Molecular Formula C5H10OS
Formula Weight 118.2
MDL Number MFCD00039937
UN Number UN3272
Beilstein 1740765
Synonym Thioacetic acid S-n-propyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2187506

  SDS
alfa aesar™
B2187506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2187514

  SDS
alfa aesar™
B2187514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2187522

  SDS
alfa aesar™
B2187522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 118.2
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
CAS 2307-10-0
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Molecular Formula C5H10OS
Chemical Name or Material S-n-Propyl thioacetate
Beilstein 1740765
UN Number UN3272
Synonym Thioacetic acid S-n-propyl ester
Assay ≥98%
Boiling Point 137° to 139°C
MDL Number MFCD00039937
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.