Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%

$134.07 - $134.07

Chemical Identifiers

Chemical Name or Material (S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine
CAS 55650-58-3
Molecular Formula C26H28FeNP
Formula Weight 441.34g/mol
MDL Number MFCD00075285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19686MC

 
alfa aesar™
L19686-MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19686ME

 
alfa aesar™
L19686-ME
500mg Each for $134.07
Description & Specifications

Specifications

Melting Point 141° to 143°C
Quantity 100mg
Optical Rotation +355° (c=0.6 in Ethanol)
Chemical Name or Material (S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine
CAS 55650-58-3
Assay 97%
Molecular Formula C26H28FeNP
Formula Weight 441.34g/mol
MDL Number MFCD00075285
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
(S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine is the raw material of asymmetric hydrogenation chiral ligands.

Solubility
Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.