missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-N,N-Dimethyl-1-phenylethylamine 98.0+%, TCI America™
$132.71 - $431.33
Chemical Identifiers
CAS | 17279-31-1 |
---|---|
Molecular Formula | C10H15N |
Molecular Weight (g/mol) | 149.237 |
MDL Number | MFCD00056236 |
InChI Key | BVURNMLGDQYNAF-VIFPVBQESA-N |
Synonym | (S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine |
PubChem CID | 2724276 |
IUPAC Name | (1S)-N,N-dimethyl-1-phenylethanamine |
SMILES | CC(C1=CC=CC=C1)N(C)C |
Chemical Identifiers
17279-31-1 | |
149.237 | |
BVURNMLGDQYNAF-VIFPVBQESA-N | |
2724276 | |
CC(C1=CC=CC=C1)N(C)C |
C10H15N | |
MFCD00056236 | |
(S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine | |
(1S)-N,N-dimethyl-1-phenylethanamine |
Specifications
17279-31-1 | |
71°C | |
MFCD00056236 | |
(S)-(-)-alpha-Methyl-N,N-dimethylbenzylamine | |
CC(C1=CC=CC=C1)N(C)C | |
149.237 | |
149.24 | |
Liquid |
Yellow | |
C10H15N | |
1 g | |
BVURNMLGDQYNAF-VIFPVBQESA-N | |
(1S)-N,N-dimethyl-1-phenylethanamine | |
2724276 | |
≥98.0% (GC) | |
(S)-(-)-N,N-Dimethyl-1-phenylethylamine |
Safety and Handling
EINECSNumber : (3)-0318
TSCA : No