Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine, 98%

$122.43 - $180.99

Chemical Identifiers

Chemical Name or Material (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
CAS 31886-57-4
Molecular Formula C14H19FeN
Formula Weight 257.16g/mol
MDL Number MFCD00066272
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1968003

 
alfa aesar™
L19680-03
1g Each for $180.99

AAL19680MD

 
alfa aesar™
L19680-MD
250mg Each for $122.43
Description & Specifications

Specifications

Boiling Point 120° to 121°C (0.7mmHg)
Quantity 1g
Flash Point 95°C (203°F)
Optical Rotation −13° (c=1 in Ethanol, 5H)
Chemical Name or Material (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
CAS 31886-57-4
Assay 98%
Molecular Formula C14H19FeN
Formula Weight 257.16g/mol
MDL Number MFCD00066272
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.