Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine, tech. 85%

$120.23 - $384.23

Chemical Identifiers

Chemical Name or Material (S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
CAS 143140-08-3
Purity Grade Technical Grade
Molecular Formula C14H25NOSi
Formula Weight 251.45
MDL Number MFCD06798960
Beilstein 5906612
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1941403

  SDS
alfa aesar™
L1941403
1g Each for $120.23

AAL1941406

  SDS
alfa aesar™
L1941406
5g Each for $384.23
Description & Specifications

Specifications

Formula Weight 251.45
Health Hazard 1 Warning
CAS 143140-08-3
Assay 85%
MDL Number MFCD06798960
Molecular Formula C14H25NOSi
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
Boiling Point 82° to 84°C (0.2mmHg)
Optical Rotation −47° (c=1 in Carbon Tetrachloride)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 5906612
Chemical Name or Material (S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.