Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride, 98+%

$94.97 - $349.42

Chemical Identifiers

Chemical Name or Material (S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride
CAS 116297-12-2
Molecular Formula C11H17N·HCl
Formula Weight 199.73
MDL Number MFCD06011753
Synonym (S)-N-(2-Propyl)-1-phenylethylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1835503

 
alfa aesar™
L1835503
1g Each for $94.97

AAL1835506

 
alfa aesar™
L1835506
5g Each for $349.42
Description & Specifications

Specifications

Formula Weight 199.73
Molecular Formula C11H17N·HCl
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
MDL Number MFCD06011753
CAS 116297-12-2
Synonym (S)-N-(2-Propyl)-1-phenylethylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 262° to 265°C
Quantity 1g
Chemical Name or Material (S)-(-)-N-Isopropyl-1-phenylethylamine hydrochloride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.