Please login to your online account to display your discounted pricing

(S)-N-FMOC-Octylglycine, 95%, 98% e.e., ACROS Organics™

$945.13 - $945.13

Chemical Identifiers

Chemical Name or Material (S)-N-FMOC-Octylglycine
Name Note 95%, (98% e.e.)
CAS 193885-59-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C25H31NO4
Formula Weight 409.52
MDL Number MFCD01311753
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC361490010

  SDS
fisher scientific acros
361490010
Glass bottle 1g Each for $945.13
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 409.52
Quantity 1g
Color White
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Infrared Spectrum Authentic
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
MDL Number MFCD01311753
Synonym (S)-N-(9-Fluorenylmethoxycarbonyl)-2-aminodecanoic acid
Chemical Name or Material (S)-N-FMOC-Octylglycine
Assay Percent Range 95%
Name Note 95%, (98% e.e.)
CAS 193885-59-5
Molecular Formula C25H31NO4