Please login to your online account to display your discounted pricing

(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-N-FMOC-α-Methylvaline
Name Note 98% ee
CAS 169566-81-8
CAS Min % 97.5
CAS Max % 100.0
CAS 1634-04-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C21H23NO4
Formula Weight 353.42
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441155000

  SDS
thermo fisher scientific
AC441155000
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441151000

  SDS
thermo fisher scientific
AC441151000
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 353.42
Quantity 500mg
Color White to pale yellow
CAS Max % 100.0
Residual Solvents 10.0% max. (MTBE)
Enantiomeric Excess 98.0% min.
Chemical Name or Material (S)-N-FMOC-α-Methylvaline
Specific Rotation +7.5° to +9.5° (20°C, 589nm) (c=1 in CHCl3)
Name Note 98% ee
Assay Percent Range 98%
Molecular Formula C21H23NO4
CAS 169566-81-8
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
CAS 1634-04-4
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym FMOC-3-Methyl-L-isovaline
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable solid.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.