Learn More
(S)-N-FMOC-α-Methylvaline, 98%, 98% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441151000
Specifications
(S)-N-FMOC-α-Methylvaline | |
Authentic | |
8.4 | |
37°C to 53°C | |
98% ee | |
97.5 | |
97.5% min. (HPLC) | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-2,3-dimethylbutanoic acid, fmoc-a-me-val-oh, fmoc-me-val-oh, fmoc-s-alpha-methylvaline, l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-d-isovaline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, s-2-fmoc-amino-2,3-dimethylbutanoic acid | |
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
353.42 | |
353.42 | |
98%,98% ee |
98.0% min. | |
10.0% max. (MTBE) | |
White to Yellow | |
100mg | |
169566-81-8,1634-04-4 | |
100.0 | |
C21H23NO4 | |
AWEZXIRZNQCCNN-NRFANRHFSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid | |
11394064 | |
Powder |
Chemical Identifiers
169566-81-8 | |
353.42 | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-2,3-dimethylbutanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-2,3-dimethylbutanoic acid, fmoc-a-me-val-oh, fmoc-me-val-oh, fmoc-s-alpha-methylvaline, l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, n-9h-fluoren-9-ylmethoxy carbonyl-3-methyl-d-isovaline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2,3-dimethylbutanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-2,3-dimethylbutanoic acid, s-2-fmoc-amino-2,3-dimethylbutanoic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2,3-dimethylbutanoic acid |
C21H23NO4 | |
AWEZXIRZNQCCNN-NRFANRHFSA-N | |
11394064 | |
CC(C)C(C)(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/equipment.
Wear protective gloves/eye protection/face protection.
In case of fire: Use carbon dioxide for extinction.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning