missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, 98% ee, ACROS Organics™

Chemical Identifiers

CAS 136552-06-2
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C19H17NO4
Formula Weight 323.35
MDL Number MFCD01320843
Synonym (S)-N-(9-Fluorenylmethoxycarbonyl)-azetidine-2-carboxylic acid
Chemical Name or Material (S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, (98% e.e.)
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC361600010 SDS Thermo Fisher Scientific
361600010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC361605000 SDS Thermo Fisher Scientific
AC361605000
Glass Bottle 500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

136552-06-2
100.0
C19H17NO4
MFCD01320843
(S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, (98% e.e.)
94.0
95%
323.35
(S)-N-(9-Fluorenylmethoxycarbonyl)-azetidine-2-carboxylic acid
≥94%

Specifications

Powder
1g
100.0
(S)-N-(9-Fluorenylmethoxycarbonyl)-azetidine-2-carboxylic acid
Glass bottle
136552-06-2
95%
(S)-N-FMOC-Azetidine-2-carboxylic acid, 95%, (98% e.e.)
323.35
White to light yellow
94.0
97.5% min. (HPLC)
Authentic
C19H17NO4
MFCD01320843
≥94%

Safety and Handling

TSCA : TSCA